8-Bromocaprylic acid
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
8-Bromooctanoic acid, ω-Bromocaprylic acid, 8-Bromooctoic acid, ω-Bromooctoic acid, ω-Bromooctanoic acid
OR16974-100G
OR16974-250G
OR16974-25G
APOSOR16974-100GEA
250
EUR
APOSOR16974-100G
APOSOR16974-250G
APOSOR16974-25G
8-Bromocaprylic acid
8-Bromocaprylic acid
Formula:
BrH₂C(CH₂)₆COOH Boiling Pt: 147…150 °C/2mmHg Melting Pt: 35…38 °C Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004430 CAS Number: 17696-11-6 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...