(Iodomethyl)cyclopentane 98%
Supplier: Apollo Scientific
Danger
OR16976-1G OR16976-250MG
APOSOR16976-1GEA 55.1 EUR
APOSOR16976-1G APOSOR16976-250MG
(Iodomethyl)cyclopentane 98%
(Iodomethyl)cyclopentane
Formula: C₆H₁₁I
Boiling Pt: 156…158 °C
Density: 1.604 g/cm³ (25 °C)
Flash Pt: 70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03453190
CAS Number: 27935-87-1
UN: 2810
ADR: 6.1,III