5,5-Dimethyl-1-pyrroline-N-oxide 97%
Supplier: Apollo Scientific
Warning
OR16990-1G OR16990-5G OR16990-10G
APOSOR16990-1GEA 29 EUR
APOSOR16990-1G APOSOR16990-5G APOSOR16990-10G
5,5-Dimethyl-1-pyrroline-N-oxide 97%
5,5-Dimethyl-1-pyrroline-N-oxide
Formula: C₆H₁₁NO
Boiling Pt: 78 °C/1.4mmHg
Melting Pt: 25…29 °C
Density: 1.015 g/cm³ (25 °C)
Flash Pt: 95 °C
Storage Temperature: Freezer
MDL Number: MFCD00005279
CAS Number: 3317-61-1
EINECS: 222-011-1