1,3-Dibromoacetone 95%
Supplier: Apollo Scientific
Danger
Synonyms: 1,3-Dibromopropan-2-one
OR16995-1G OR16995-5G OR16995-10G
APOSOR16995-1GEA 21.9 EUR
APOSOR16995-1G APOSOR16995-5G APOSOR16995-10G
1,3-Dibromoacetone 95%
1,3-Dibromoacetone
Formula: BrH₂CC(O)CH₂Br
Boiling Pt: 97…98 °C/21mmHg
Density: 2.12 g/cm³ (25 °C)
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00013540
CAS Number: 816-39-7
EINECS: 212-430-8
UN: 1760
ADR: 8,II