5-Bromo-4-chloro-7H-pyrrolo[2,3-d]pyrimidine
Supplier: Apollo Scientific
Danger
Synonyms: 7-Bromo-6-chloro-7-deazapurine
OR17052-1G OR17052-5G
APOSOR17052-1GEA 21.9 EUR
APOSOR17052-1G APOSOR17052-5G
5-Bromo-4-chloro-7H-pyrrolo[2,3-d]pyrimidine
5-Bromo-4-chloro-7H-pyrrolo[2,3-d]pyrimidine
Formula: C₆H₃BrClN₃
MW: 232.47 g/mol
Melting Pt: 221…225 °C
Storage Temperature: Ambient
MDL Number: MFCD09702029
CAS Number: 22276-95-5
UN: 2811
ADR: 6.1 III