3-Bromo-5-nitropyridin-4-amine 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Amino-3-bromo-5-nitropyridine
OR17112-1G OR17112-25G OR17112-5G
APOSOR17112-1GEA 21.9 EUR
APOSOR17112-1G APOSOR17112-25G APOSOR17112-5G
3-Bromo-5-nitropyridin-4-amine 97%
3-Bromo-5-nitropyridin-4-amine
Formula: C5H4BrN3O2
MW: 218.0100 g/mol
Melting Pt: 179…181 °C
Storage Temperature: Ambient
MDL Number: MFCD01646057
CAS Number: 89284-05-9