2-[(Acetylthio)methyl]-3-phenylpropionic acid 96%
Supplier: Apollo Scientific
Warning
Synonyms: 2-[(Acetylthio)methyl]-3-phenylpropionic Acid
OR17540-5G OR17540-25G OR17540-100G
APOSOR17540-5GEA 21.9 EUR
APOSOR17540-5G APOSOR17540-25G APOSOR17540-100G
2-[(Acetylthio)methyl]-3-phenylpropionic acid 96%
2-[(Acetylthio)methyl]-3-phenylpropionic acid
Formula: C₁₂H₁₄O₃S
MW: 238.30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236751
CAS Number: 91702-98-6