2-Oxa-6-azaspiro[3.5]nonane 95%
Supplier: Apollo Scientific
Warning
OR17716-250MG OR17716-100MG
APOSOR17716-250MGEA 426 EUR
APOSOR17716-250MG APOSOR17716-100MG
2-Oxa-6-azaspiro[3.5]nonane 95%
2-Oxa-6-azaspiro[3.5]nonane
Formula: C₇H₁₃NO
Boiling Pt: 208 °C
Density: 1.04 g/cm³ (25 °C)
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD12407095
CAS Number: 1046153-20-1