2-Chloroethyl methyl ether
Supplier: Apollo Scientific
Danger
OR17824-100G OR17824-25G OR17824-500G
APOSOR17824-100GEA 39.3 EUR
APOSOR17824-100G APOSOR17824-25G APOSOR17824-500G
2-Chloroethyl methyl ether
2-Chloroethyl methyl ether
Formula: C₃H₇ClO
Boiling Pt: 89…90 °C
Melting Pt: –55 °C
Density: 1.035 g/cm³ (25 °C)
Flash Pt: 10 °C
Storage Temperature: Ambient
MDL Number: MFCD00000972
CAS Number: 627-42-9
EINECS: 210-999-7
UN: 3271
ADR: 3,II