(2-Methylpyrimidin-5-yl)methanol 98%
Supplier: Apollo Scientific
Warning
OR17837-1G OR17837-5G OR17837-25G
APOSOR17837-1GEA 106 EUR
APOSOR17837-1G APOSOR17837-5G APOSOR17837-25G
(2-Methylpyrimidin-5-yl)methanol 98%
(2-Methylpyrimidin-5-yl)methanol
Formula: C₆H₈N₂O
Boiling Pt: 237 °C
Melting Pt: 102…103 °C
Density: 1.171 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD09414732
CAS Number: 2239-83-0