Bis(2-iodoethyl) ether 97%
Supplier: Apollo Scientific
Danger
Synonyms: 2,2'-Diiododiethyl ether, 1-Iodo-2-(2-iodoethoxy)ethane, 2-Iodoethyl ether
OR18271-1G OR18271-25G OR18271-5G
APOSOR18271-1GEA 62.5 EUR
APOSOR18271-1G APOSOR18271-25G APOSOR18271-5G
Bis(2-iodoethyl) ether 97%
Bis(2-iodoethyl) ether
Formula: C₄H₈I₂O
MW: 325.92 g/mol
Boiling Pt: 123.5…124 °C/10mmHg
Density: 2.296 g/cm³ (25 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00092250
CAS Number: 34270-90-1