2-Amino-6-methoxybenzothiazole 98%
Supplier: Apollo Scientific
Warning
Synonyms: 6-Methoxybenzothiazol-2-ylamine
OR18278-100G OR18278-500G OR18278-1KG
APOSOR18278-100GEA 21.9 EUR
APOSOR18278-100G APOSOR18278-500G APOSOR18278-1KG
2-Amino-6-methoxybenzothiazole 98%
2-Amino-6-methoxybenzothiazole
Formula: C₈H₈N₂OS
MW: 180.23 g/mol
Boiling Pt: 348
Melting Pt: 161…167 °C
Storage Temperature: Ambient
MDL Number: MFCD00005787
CAS Number: 1747-60-0
EINECS: 217-130-0