(Isopropylthio)benzene 97%
Supplier: Apollo Scientific
Warning
Synonyms: Isopropyl phenyl sulphide
OR18312-1KG OR18312-250G OR18312-50G
APOSOR18312-1KGEA 122 EUR
APOSOR18312-1KG APOSOR18312-250G APOSOR18312-50G
(Isopropylthio)benzene 97%
(Isopropylthio)benzene
Formula: C₉H₁₂S
MW: 152.26 g/mol
Boiling Pt: 105 °C/ 40mmHg
Melting Pt: 56
Density: 0.9852 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00039842
CAS Number: 3019-20-3
EINECS: 221-162-0