1-Bromo-2-iodobenzene 98+%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromo-1-iodobenzene, 2-Bromoiodobenzene
OR18313-100G OR18313-25G
APOSOR18313-100GEA 40.6 EUR
APOSOR18313-100G APOSOR18313-25G
1-Bromo-2-iodobenzene 98+%
1-Bromo-2-iodobenzene
Formula: C₆H₄BrI
Boiling Pt: 257 °C
Melting Pt: 44478 °C
Density: 2.203 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001030
CAS Number: 583-55-1
EINECS: 209-508-9