1,4-Diethoxybenzene 98%
Supplier: Apollo Scientific
Warning
OR18328-100G OR18328-25G OR18328-500G
APOSOR18328-100GEA 116 EUR
APOSOR18328-100G APOSOR18328-25G APOSOR18328-500G
1,4-Diethoxybenzene 98%
1,4-Diethoxybenzene
Formula: C₁₀H₁₄O₂
Boiling Pt: 245…248 °C
Melting Pt: 69…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00015146
CAS Number: 122-95-2
EINECS: 204-585-5