Fluorene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 9H-Fluorene, Diphenylenmethane
OR18343-100G OR18343-250G OR18343-500G
APOSOR18343-100GEA 39.3 EUR
APOSOR18343-100G APOSOR18343-250G APOSOR18343-500G
Fluorene 98%
Fluorene
Formula: C₁₃H₁₀
MW: 166.22 g/mol
Boiling Pt: 295 °C
Melting Pt: 112…115 °C
Density: 1.202 g/cm³ (25 °C)
Flash Pt: 151 °C
Storage Temperature: Ambient
MDL Number: MFCD00001111
CAS Number: 86-73-7
EINECS: 201-695-5
UN: 3077
ADR: 9+EHS III
Merck Index: 13,04181