5-Bromothiophene-2-carbaldehyde 97%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Bromo-2-thiophenecarboxaldehyde
OR18347-100G OR18347-25G OR18347-1G
APOSOR18347-100GEA 144 EUR
APOSOR18347-100G APOSOR18347-25G APOSOR18347-1G
5-Bromothiophene-2-carbaldehyde 97%
5-Bromothiophene-2-carbaldehyde
Formula: C₅H₃BrOS
Boiling Pt: 105…107 °C/11mmHg
Melting Pt: 15 °C
Density: 1.607 g/cm³ (25 °C)
Flash Pt: 99 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005432
CAS Number: 4701-17-1
EINECS: 225-176-8