4-Benzoylbenzoic acid
Supplier: Apollo Scientific
Warning
OR18378-25G OR18378-5G
APOSOR18378-25GEA 150 EUR
APOSOR18378-25G APOSOR18378-5G
4-Benzoylbenzoic acid
4-Benzoylbenzoic acid
Formula: C₁₄H₁₀O₃
Melting Pt: 198…200 °C
Storage Temperature: Ambient
MDL Number: MFCD00002560
CAS Number: 611-95-0
EINECS: 210-286-0