1,3,5-Tris(bromomethyl)benzene 97+%
Supplier: Apollo Scientific
Danger
OR18429-1G OR18429-5G OR18429-25G
APOSOR18429-1GEA 21.9 EUR
APOSOR18429-1G APOSOR18429-5G APOSOR18429-25G
1,3,5-Tris(bromomethyl)benzene 97+%
1,3,5-Tris(bromomethyl)benzene
Formula: C₉H₉Br₃
MW: 356.88 g/mol
Melting Pt: 94…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00182500
CAS Number: 18226-42-1
UN: 3261
ADR: 8 II