Sulphamoyl chloride
Supplier: Apollo Scientific
Danger
Synonyms: Sulphamoyl Chloride
OR18738-5G OR18738-1G
APOSOR18738-5GEA 27.7 EUR
APOSOR18738-5G APOSOR18738-1G
Sulphamoyl chloride
Sulphamoyl chloride
Formula: ClH₂NO₂S
Melting Pt: 40 °C
Storage Temperature: Freezer
MDL Number: MFCD11519990
CAS Number: 7778-42-9
UN: 3261
ADR: 8,II