p-Tolyl chloroformate

Supplier: Apollo Scientific
Danger

Synonyms: 4-Methylphenyl chloroformate

OR1994-1G OR1994-500MG OR1994-5G
APOSOR1994-1GEA 58 EUR
APOSOR1994-1G APOSOR1994-500MG APOSOR1994-5G
p-Tolyl chloroformate
p-Tolyl chloroformate
Formula: C₈H₇ClO₂
Boiling Pt: 99.5 °C/20mmHg
Density: 1.188 g/cm³ (25 °C)
Flash Pt: 86 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013255
CAS Number: 937-62-2
EINECS: 213-332-8
UN: 3277
ADR: 6.1+8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR