(2,5-Dimethoxy-2,5-dihydrofuran-2-yl)methylamine 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-(Aminomethyl)-2,5-dihydro-2,5-dimethoxyfuran
OR20015-1G OR20015-5G OR20015-10G
APOSOR20015-1GEA 84.1 EUR
APOSOR20015-1G APOSOR20015-5G APOSOR20015-10G
(2,5-Dimethoxy-2,5-dihydrofuran-2-yl)methylamine 97%
(2,5-Dimethoxy-2,5-dihydrofuran-2-yl)methylamine
Formula: C₇H₁₃NO₃
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00082196
CAS Number: 14496-27-6