5-Chloro-2-iodopyrimidine 97%

Supplier: Apollo Scientific
Warning

OR20045-100G OR20045-5G OR20045-25G
APOSOR20045-100GEA 847 EUR
APOSOR20045-100G APOSOR20045-5G APOSOR20045-25G
5-Chloro-2-iodopyrimidine 97%
5-Chloro-2-iodopyrimidine
Formula: C₄H₂ClIN₂
Storage Temperature: Freezer
MDL Number: MFCD08062390
CAS Number: 874676-81-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR