4-(3-Pyridyl)aniline
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Pyridin-3-ylaniline
OR2020-1G
OR2020-250MG
APOSOR2020-1GEA
393
EUR
APOSOR2020-1G
APOSOR2020-250MG
4-(3-Pyridyl)aniline
4-(3-Pyridyl)aniline
Formula:
C₁₁H₁₀N₂ Melting Pt: 115…117 °C Storage Temperature: Ambient |
MDL Number:
MFCD00956763 CAS Number: 82261-42-5 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...