Methyl nitroacetate

Supplier: Apollo Scientific
Warning

OR2038-1G OR2038-25G OR2038-5G
APOSOR2038-1GEA 21.9 EUR
APOSOR2038-1G APOSOR2038-25G APOSOR2038-5G
Methyl nitroacetate
Formula: C₃H₅NO₄
Boiling Pt: 195…198 °C
Density: 1.294 g/cm³ (25 °C)
Flash Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00075480
CAS Number: 2483-57-0
EINECS: 219-622-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR