trans-3-Chloroacrylic acid 99%
Supplier: Apollo Scientific
Danger
OR2048-1G OR2048-5G
APOSOR2048-1GEA 148 EUR
APOSOR2048-1G APOSOR2048-5G
trans-3-Chloroacrylic acid 99%
trans-3-Chloroacrylic acid
Formula: C₃H₃ClO₂
Melting Pt: 81…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00064237
CAS Number: 2345-61-1
EINECS: 219-070-0
UN: 3261
ADR: 8,II