2,3-Dimethylmaleic anhydride 98%
Supplier: Apollo Scientific
Warning
Synonyms: 3,4-Dimethylfuran-2,5-dione
OR21481-100G OR21481-25G OR21481-5G
APOSOR21481-100GEA 60.9 EUR
APOSOR21481-100G APOSOR21481-25G APOSOR21481-5G
2,3-Dimethylmaleic anhydride 98%
2,3-Dimethylmaleic anhydride
Formula: C₆H₆O₃
MW: 126.11 g/mol
Boiling Pt: 222…223 °C
Melting Pt: 93…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00005523
CAS Number: 766-39-2
EINECS: 212-165-8