Pyrazine-2,3-dicarboxylic acid 99%
Supplier: Apollo Scientific
Warning
OR21608-25G OR21608-100G
APOSOR21608-25GEA 21.9 EUR
APOSOR21608-25G APOSOR21608-100G
Pyrazine-2,3-dicarboxylic acid 99%
Pyrazine-2,3-dicarboxylic acid
Formula: C₆H₄N₂O₄
MW: 168.11 g/mol
Melting Pt: 188 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006131
CAS Number: 89-01-0
EINECS: 201-875-3