1-Chlorophthalazin-4-one 96%
Supplier: Apollo Scientific
Warning
OR21968-1G OR21968-250MG OR21968-5G
APOSOR21968-1GEA 40.6 EUR
APOSOR21968-1G APOSOR21968-250MG APOSOR21968-5G
1-Chlorophthalazin-4-one 96%
1-Chlorophthalazin-4-one
Formula: C₈H₅ClN₂O
Melting Pt: 270…274 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00102192
CAS Number: 2257-69-4