Methyl-3-bromobenzoate
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 3-bromobenzoate, 3-Bromobenzoic acid methyl ester
OR2197-100G OR2197-25G
APOSOR2197-100GEA 111 EUR
APOSOR2197-100G APOSOR2197-25G
Methyl-3-bromobenzoate
Methyl 3-bromobenzoate
Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Boiling Pt: 127…128 °C
Melting Pt: 31…33 °C
Storage Temperature: Ambient
MDL Number: MFCD00017777
CAS Number: 618-89-3