4-Ethylphenylacetylene 97%
Supplier: Apollo Scientific
Warning
Synonyms: (4-Ethylphenyl)acetylene, 1-Ethyl-4-ethynylbenzene, 1-Ethynyl-4-ethylbenzene, (4-Ethylphenyl)ethyne
OR2201-100G OR2201-25G OR2201-5G
APOSOR2201-100GEA 171 EUR
APOSOR2201-100G APOSOR2201-25G APOSOR2201-5G
4-Ethylphenylacetylene 97%
4-Ethylphenylacetylene
Formula: C₁₀H₁₀
Boiling Pt: 30 °C/0.05mmHg
Density: 0.93 g/cm³ (25 °C)
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00173887
CAS Number: 40307-11-7
UN: 3334
ADR: 9,III