(±)-Ethyl-2-bromopropionate
Supplier: Apollo Scientific
Danger
Synonyms: DL-Ethyl 2-bromopropionate, Ethyl 2-bromopropionate
OR2205-10KG OR2205-2.5KG OR2205-500G
APOSOR2205-10KGEA 522 EUR
APOSOR2205-10KG APOSOR2205-2.5KG APOSOR2205-500G
(±)-Ethyl-2-bromopropionate
(±)-Ethyl 2-bromopropionate
Formula: C₅H₉BrO₂
MW: 181.03 g/mol
Boiling Pt: 156…160 °C
Density: 1.394 g/cm³ (25 °C)
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00000144
CAS Number: 535-11-5
EINECS: 208-609-2
UN: 2920
ADR: 8+3 II