3,5-Di-tert-butylpyrocatechol
Supplier: Apollo Scientific
Warning
OR22089-100G OR22089-25G
APOSOR22089-100GEA 192 EUR
APOSOR22089-100G APOSOR22089-25G
3,5-Di-tert-butylpyrocatechol
3,5-Di-tert-butylpyrocatechol
Formula: C₁₄H₂₂O₂
Boiling Pt: 140…145 °C/5mmHg
Melting Pt: 95…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00008819
CAS Number: 1020-31-1