4-Ethylbenzoyl chloride
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
4-Ethylbenzene-1-carbonyl chloride, 4-Ethylbenzoylchloride
OR22097-10G
OR22097-50G
APOSOR22097-10GEA
126
EUR
APOSOR22097-10G
APOSOR22097-50G
4-Ethylbenzoyl chloride
4-Ethylbenzoyl chloride
Formula:
C₉H₉ClO Storage Temperature: Ambient |
MDL Number:
MFCD00000697 CAS Number: 16331-45-6 EINECS: 240-404-6 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...