Cyclododecanone
Supplier: Apollo Scientific
OR22155-100G OR22155-25G
APOSOR22155-100GEA 162 EUR
APOSOR22155-100G APOSOR22155-25G
Cyclododecanone
Cyclododecanone
Formula: C₁₂H₂₂O
Boiling Pt: 277 °C/760mmHg
Melting Pt: 59…61 °C
Flash Pt: 118 °C
Storage Temperature: Ambient
MDL Number: MFCD00003722
CAS Number: 830-13-7
UN: 3077
ADR: 9+EHS,III