4-(1,3,4-Oxadiazol-2-yl)aniline
Supplier: Apollo Scientific
Warning
OR2220-1G OR2220-100MG
APOSOR2220-1GEA 125 EUR
APOSOR2220-1G APOSOR2220-100MG
4-(1,3,4-Oxadiazol-2-yl)aniline
4-(1,3,4-Oxadiazol-2-yl)aniline
Formula: C₈H₇N₃O
Melting Pt: 137…139 °C
Storage Temperature: Ambient
MDL Number: MFCD08059433
CAS Number: 35219-13-7