2,4-Dihydroxybenzophenone 99%
Supplier: Apollo Scientific
Warning
OR2307-500G OR2307-100G
APOSOR2307-500GEA 57.2 EUR
APOSOR2307-500G APOSOR2307-100G
2,4-Dihydroxybenzophenone 99%
2,4-Dihydroxybenzophenone
Formula: C₁₃H₁₀O₃
Melting Pt: 144…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00002277
CAS Number: 131-56-6