2-Quinolinecarbaldehyde
Supplier: Apollo Scientific
Warning
OR23073-25G OR23073-5G
APOSOR23073-25GEA 0 EUR
APOSOR23073-25G APOSOR23073-5G
2-Quinolinecarbaldehyde
2-Quinolinecarbaldehyde
Formula: C₁₀H₇NO
Melting Pt: 70…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00075032
CAS Number: 5470-96-2
EINECS: 226-804-3