6-Bromoquinoline
Supplier: Apollo Scientific
Danger
OR23076-10G OR23076-250G OR23076-50G
APOSOR23076-10GEA 21.9 EUR
APOSOR23076-10G APOSOR23076-250G APOSOR23076-50G
6-Bromoquinoline
6-Bromoquinoline
Formula: C₉H₆BrN
MW: 208.06
Melting Pt: 24 °C
Density: 1.538 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00024023
CAS Number: 5332-25-2