5-Bromo-m-xylene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Bromo-3,5-dimethylbenzene
OR2316-100G OR2316-1KG OR2316-500G
APOSOR2316-100GEA 33.5 EUR
APOSOR2316-100G APOSOR2316-1KG APOSOR2316-500G
5-Bromo-m-xylene 98%
5-Bromo-m-xylene
Formula: C₈H₉Br
Boiling Pt: 202…204 °C
Density: 1.361 g/cm³ (25 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00000087
CAS Number: 556-96-7