2-Morpholino-3-pyridinamine
Supplier: Apollo Scientific
Danger
Synonyms: 3-Amino-2-morpholin-4-ylpyridine
OR23393-10G OR23393-1G OR23393-100MG
APOSOR23393-10GEA 377 EUR
APOSOR23393-10G APOSOR23393-1G APOSOR23393-100MG
2-Morpholino-3-pyridinamine
2-Morpholino-3-pyridinamine
Formula: C₉H₁₃N₃O
Melting Pt: 127…128 °C
Storage Temperature: Ambient
MDL Number: MFCD03001243
CAS Number: 51627-47-5
UN: 2811
ADR: 6.1,III