5-Bromobenzo[b]thiophene 98%
Supplier: Apollo Scientific
|
Warning
|
OR23397-5G
OR23397-25G
APOSOR23397-5GEA
81.2
EUR
APOSOR23397-5G
APOSOR23397-25G
5-Bromobenzo[b]thiophene 98%
5-Bromobenzo[b]thiophene
Formula:
C₈H₅BrS Boiling Pt: 105…107 °C/10mmHg Melting Pt: 44…48 °C Storage Temperature: Ambient |
MDL Number:
MFCD03069318 CAS Number: 4923-87-9 |