3,5-Dichlorobenzonitrile 97%
Supplier: Apollo Scientific
Warning
OR23753-100G OR23753-10G OR23753-5G
APOSOR23753-100GEA 81.2 EUR
APOSOR23753-100G APOSOR23753-10G APOSOR23753-5G
3,5-Dichlorobenzonitrile 97%
3,5-Dichlorobenzonitrile
Formula: C₇H₃Cl₂N
MW: 172.01 g/mol
Boiling Pt: 269
Melting Pt: 64…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00001800
CAS Number: 6575-00-4
EINECS: 229-495-3
UN: 3439
ADR: 6.1 III