4-(Dichloromethyl)benzonitrile
Supplier: Apollo Scientific
Danger
OR23830-1G OR23830-5G
APOSOR23830-1GEA 368 EUR
APOSOR23830-1G APOSOR23830-5G
4-(Dichloromethyl)benzonitrile
4-(Dichloromethyl)benzonitrile
Formula: C₈H₅Cl₂N
MW: 186.0400 g/mol
Boiling Pt: 273…276 °C
Melting Pt: 44 °C
Storage Temperature: Ambient
MDL Number: MFCD00045606
CAS Number: 74231-65-5
UN: 3439
ADR: 6.1 III