4-Chlorostyrene
Supplier: Apollo Scientific
Warning
OR2398-100G OR2398-1KG OR2398-250G
APOSOR2398-100GEA 72.5 EUR
APOSOR2398-100G APOSOR2398-1KG APOSOR2398-250G
4-Chlorostyrene
4-Chlorostyrene
Formula: C₈H₇Cl
Boiling Pt: 192 °C
Melting Pt: –16 °C
Density: 1.155 g/cm³ (25 °C)
Flash Pt: 68 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000632
CAS Number: 1073-67-2
EINECS: 214-028-8