tert-Butyl (2-hydrazinyl-2-oxoethyl)carbamate
Supplier: Apollo Scientific
Warning
Synonyms: 2-Aminoacetohydrazide, 2-BOC protected
OR2444-1G OR2444-5G
APOSOR2444-1GEA 21.9 EUR
APOSOR2444-1G APOSOR2444-5G
tert-Butyl (2-hydrazinyl-2-oxoethyl)carbamate
tert-Butyl (2-hydrazinyl-2-oxoethyl)carbamate
Formula: C₇H₁₅N₃O₃
MW: 189.2100 g/mol
Melting Pt: 111…115 °C
Storage Temperature: Ambient
MDL Number: MFCD01570558
CAS Number: 6926-09-6