Ethyl-4-(dimethylamino)-2-oxobut-3-enoate
Supplier: Apollo Scientific
Warning
OR24503-1G OR24503-250MG
APOSOR24503-1GEA 279 EUR
APOSOR24503-1G APOSOR24503-250MG
Ethyl-4-(dimethylamino)-2-oxobut-3-enoate
Ethyl-4-(dimethylamino)-2-oxobut-3-enoate
Formula: C₈H₁₃NO₃
Boiling Pt: 180…185 °C/0.7mmHg
Melting Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00219095
CAS Number: 67751-14-8