Pyridino[3,2-b]pyrazine
Supplier: Apollo Scientific
Synonyms: Pyrido[2,3-b]pyrazine
OR24673-1G OR24673-250MG OR24673-500MG
APOSOR24673-1GEA 97.3 EUR
APOSOR24673-1G APOSOR24673-250MG APOSOR24673-500MG
Pyridino[3,2-b]pyrazine
Pyridino[3,2-b]pyrazine
Formula: C₇H₅N₃
MW: 131.14 g/mol
Melting Pt: 142…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00006717
CAS Number: 322-46-3