1-Methyl-1,5-dihydro-4H-pyrazolo[3,4-d]pyrimidin-4-one
Supplier: Apollo Scientific
Warning
OR2482-100MG OR2482-1G OR2482-5G
APOSOR2482-100MGEA 21.9 EUR
APOSOR2482-100MG APOSOR2482-1G APOSOR2482-5G
1-Methyl-1,5-dihydro-4H-pyrazolo[3,4-d]pyrimidin-4-one
1-Methyl-1,5-dihydro-4H-pyrazolo[3,4-d]pyrimidin-4-one
Formula: C₆H₆N₄O
Melting Pt: 290…292 °C
Storage Temperature: Ambient
MDL Number: MFCD01312343
CAS Number: 5334-56-5