2-(4-Chlorophenyl)ethanol 99%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Chlorophenethyl alcohol
OR24970-100G OR24970-25G
APOSOR24970-100GEA 33.5 EUR
APOSOR24970-100G APOSOR24970-25G
2-(4-Chlorophenyl)ethanol 99%
2-(4-Chlorophenyl)ethanol
Formula: C₈H₉ClO
MW: 156.61 g/mol
Boiling Pt: 110 °C/0.5mmHg
Density: 1.157 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002899
CAS Number: 1875-88-3
EINECS: 217-506-4